missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(acrylamide), carboxyl modified, pure, low carboxyl content, ∽ MW 200,000, Thermo Scientific™
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Specifications
Color | White |
Physical Form | Powder or Granules |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol |
Quantity | 5 g |
Formula Weight | ≈200.000 |
Grade | Pure |
Infrared Spectrum | Authentic |
Loss on Drying | 10% max. |
Packaging | Glass bottle |
Chemical Name or Material | Poly(acrylamide), carboxyl modified, Carboxyl Modified, Low Carboxyl Conten |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'