missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol |
Chemical Name or Material | Poly(acrylamide), carboxyl modified |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'