missing translation for 'onlineSavingsMsg'
Learn More

Poly(4-chlorostyrene), Thermo Scientific™

Catalog Number 10644682
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 24991-47-7
Molecular Formula C8H7Cl
Linear Formula [-CH2CH(C6H4Cl)-]n
MDL Number MFCD00084381
Density 1.5500g/mL
Synonym 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
Quantity 5g
InChI Key KTZVZZJJVJQZHV-UHFFFAOYSA-N
SMILES C=CC1=CC=C(C=C1)Cl
IUPAC Name 1-chloro-4-ethenylbenzene
Molecular Weight (g/mol) 138.594
PubChem CID 14085
Formula Weight 51000
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass bottle
Specific Gravity 1.55
Chemical Name or Material Poly(4-chlorostyrene)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'