missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-octylthiophene-2,5-diyl), regiorandom, low metals, Thermo Scientific Chemicals

Catalog Number p-7059524
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
104934-51-2
(C12H18S)n
241.44
MFCD00241412
RXIQSINDOTYIGV-UHFFFAOYSA-N
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
121233643
azane;2,5-dimethyl-3-octylthiophene
CCCCCCCCC1=C(-*)SC(-*)=C1
This item is not returnable. View return policy
104934-51-2
(C12H18S)n
241.44
MFCD00241412
RXIQSINDOTYIGV-UHFFFAOYSA-N
poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
121233643
azane;2,5-dimethyl-3-octylthiophene
CCCCCCCCC1=C(-*)SC(-*)=C1

Conducting polymers

Conducting polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Solubility Information Soluble in THF, CHCl3
Quantity 0.25 g
Chemical Name or Material Poly (3-octylthiophene-2,5-diyl), Regiorandom, low metals

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'