missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals

Catalog Number 15462547
Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy
104934-50-1
(C10H14S)n
NaN
MFCD00217686
DUFPJSOXRHVDOV-UHFFFAOYSA-N
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
22950249
3-hexyl-2,5-dimethylthiophene
CCCCCCC1=C(-*)SC(-*)=C1
This item is not returnable. View return policy
104934-50-1
(C10H14S)n
NaN
MFCD00217686
DUFPJSOXRHVDOV-UHFFFAOYSA-N
poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
22950249
3-hexyl-2,5-dimethylthiophene
CCCCCCC1=C(-*)SC(-*)=C1

Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) research and devices. It is also used in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Physical Form Powder/Chunks
Solubility Information Soluble (50mg/ml) in chlorinated solvents such as chloroform,chlorobenzene,dichlorobenzene and trichlorobenzene.
Quantity 2 g
Sensitivity Light sensitive
Chemical Name or Material Poly (3-hexylthiophene-2,5-diyl)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'