missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-butylthiophene-2,5-diyl), regiorandom, Thermo Scientific™

Catalog Number p-7059515
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
98837-51-5
(C8H10S)n
168.30
MFCD00677712
DUOSBQJOYVIVOR-UHFFFAOYSA-N
poly 3-butylthiophene-2,5-diyl , regiorandom, poly 3-butylthiophene-2,5-diyl , regioregular
23533119
3-butyl-2,5-dimethylthiophene
CCCCC1=C(-*)SC(-*)=C1
This item is not returnable. View return policy
98837-51-5
(C8H10S)n
168.30
MFCD00677712
DUOSBQJOYVIVOR-UHFFFAOYSA-N
poly 3-butylthiophene-2,5-diyl , regiorandom, poly 3-butylthiophene-2,5-diyl , regioregular
23533119
3-butyl-2,5-dimethylthiophene
CCCCC1=C(-*)SC(-*)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown to Black
Physical Form Powder
Quantity 0.25 g
Chemical Name or Material Poly (3-butylthiophene-2,5-diyl), Regiorandom
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'