missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, Thermo Scientific Chemicals

Catalog Number p-3779908
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
27119-07-9, 7732-18-5
(C7H13NO4S)n
207.24
MFCD00084369
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*
This item is not returnable. View return policy
27119-07-9, 7732-18-5
(C7H13NO4S)n
207.24
MFCD00084369
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Viscous Liquid
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
Quantity 25 g
Percent Purity 8.0 to 12.0% (solids)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution
compliance-icons
Product Identifier
  • Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'