missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565733
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LALKQELRETM KRMPYFIETP EERQDIERYS KRYMKVYKEE WIPDWR Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3G is a specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It may direct with other members of the RPC3/POLR3C-RPC6/POLR3F-RPC7/POLR3G subcomplex RNA Pol III binding to the TFIIIB-DNA complex via the interactions between TFIIIB and POLR3F. It may be involved either in the recruitment and stabilization of the subcomplex within RNA polymerase III, or in stimulating catalytic functions of other subunits during initiation. POLR3G plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. POLR3G acts as nuclear and cytosolic DNA sensor involved in innate immune response. POLR3G can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.Specifications
POLR3G | |
Polyclonal | |
Unconjugated | |
POLR3G | |
2310047G20Rik; AV275904; DNA-directed RNA polymerase III 32 kDa polypeptide; DNA-directed RNA polymerase III subunit G; DNA-directed RNA polymerase III subunit RPC7; Polr3g; polymerase (RNA) III (DNA directed) polypeptide G; polymerase (RNA) III (DNA directed) polypeptide G (32kD); polymerase (RNA) III subunit G; RNA polymerase III 32 kDa apha subunit; RNA polymerase III 32 kDa subunit; RNA polymerase III subunit C7; RNA polymerase III subunit G; RPC32; RPC32-alpha; RPC7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10622 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15318 | |
POLR3G | |
Recombinant Human POLR3G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only