Learn More
POLR2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565128
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: NRETIASVRSL LLQKKLHKFE LACLANLCPE TAEESKALIP SLEGRFEDEE LQQILDDIQT KRSFQY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3' untranslated region of the transcript from this gene.Specifications
POLR2D | |
Polyclonal | |
Unconjugated | |
Polr2d | |
2310002G05Rik; 2610028L19Rik; DNA-directed RNA polymerase II 16 kDa polypeptide; DNA-directed RNA polymerase II subunit D; DNA-directed RNA polymerase II subunit RPB4; HSRBP4; HSRPB4; plasma; POLR2D; polymerase (RNA) II (DNA directed) polypeptide D; polymerase (RNA) II subunit D; RBP4; RNA polymerase II 16 kDa subunit; RNA polymerase II subunit B4; RNA polymerase II subunit D; RNA polymerase II subunit hsRBP4; RPB16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15514 | |
Polr2d | |
Recombinant Human POLR2D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only