missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poloxamer 188, Thermo Scientific Chemicals
90.90 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.
67.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21648" to get your promotional price.
67.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Description
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
Solubility
Miscible with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
Solubility
Miscible with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
CAS | 9003-11-6 |
Color | White |
Content And Storage | Ambient temperatures |
Quantity | 500 g |
Solubility | Miscible with water. |
Form | Solid |
Chemical Name or Material | Poloxamer 188 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'