Learn More
SCHOTT AG Lighting and Imaging™ Twistable Polarization Filter for Focusing Lens
This Twistable Polarization Filter for Focusing Lens allows better viewing of polymeric structure properties in many materials. For use with focusing lens 158 215.
Brand:  SCHOTT AG Lighting and Imaging™ 158206
82.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10045671
  • £89.60 / Each
Only null left
Estimated Shipment: 18-09-2025
Add to basket
Description
Description
  • Polarization filter.
  • Twistable.
  • Polarized light makes properties of the materials visible (polymeric structures).
  • For use with focusing lens 158215.
Specifications
Specifications
Polarization Filter
Stereo Microscopy Illumination
Focusing Lens 158215