Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter for Focusing Lens
The Polarization Filter for Focusing Lens allows better viewing of polymeric structure properties in many materials. For use with focusing lens 158 210.
Brand: SCHOTT AG Lighting and Imaging™ 158205
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Polarization filter.
- Polarized light makes properties of the materials visible (polymeric structures).
- For use with focusing lens 158210.
Specifications
Polarization Filter | |
Model: KL 1600 LED, KL 2500 LED, KL 1500 HAL, KL300LED |