missing translation for 'onlineSavingsMsg'
Learn More

Podoplanin, Biotin, clone: eBio8.1.1 (8.1.1), eBioscience™

Syrian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5381-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279829

  • £49.20 / 25Āµg


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

The 8.1.1 monoclonal antibody reacts with mouse podoplanin (T1a, gp38, aggrus), a 43 kDa transmembrane glycoprotein, named for its expression in kidney glomerular epithelial cells (podocytes). In addition, Podoplanin is expressed in epithelial and mesothelial cells such as intestinal epithelium, alveolar type I cells, podocytes, and mesothelium of the visceral peritoneum. It was also shown to be a potent marker for lymphatic endothelium. Podoplanin is also expressed by subcapsular epithelial cells of the murine thymus. Mice deficient in Podoplanin die at birth because of a respiratory defect and congenital lymphedema due to a failure in lymphatic pattern formation.
Specifications

Specifications

Podoplanin
eBio8.1.1 (8.1.1)
Biotin
Conjugated
Golden Syrian Hamster
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow CytometryĆ¾Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Pdpn
IgG
RUO
Primary
Murine
Videos

For Research Use Only.