missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Podophyllotoxin, 95%

Catalog Number p-185568
32.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
518-28-5
C22H22O8
414.41
MFCD00075290
YJGVMLPVUAXIQN-XVVDYKMHSA-N
podophyllotoxin, podofilox, condylox, condyline, wartec, podophyllinic acid lactone, podophyllotoxin 7, --podophyllotoxin, warticon, podophyllum
10607
CHEBI:50305
(5R,5aR,8aR,9R)-5-hydroxy-9-(3,4,5-trimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
COC1=CC(=CC(OC)=C1OC)[C@H]1[C@@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
518-28-5
C22H22O8
414.41
MFCD00075290
YJGVMLPVUAXIQN-XVVDYKMHSA-N
podophyllotoxin, podofilox, condylox, condyline, wartec, podophyllinic acid lactone, podophyllotoxin 7, --podophyllotoxin, warticon, podophyllum
10607
CHEBI:50305
(5R,5aR,8aR,9R)-5-hydroxy-9-(3,4,5-trimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
COC1=CC(=CC(OC)=C1OC)[C@H]1[C@@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178.0°C to 184.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 7663
Specific Rotation − 110.70
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone and acetic acid
Formula Weight 414.4
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Podophyllotoxin, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Podophyllotoxin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'