missing translation for 'onlineSavingsMsg'
Learn More

Podocalyxin Mouse anti-Human, PE, Clone: B34D1.3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-8873-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15597136

  • £79.25 / 25 tests

Estimated Shipment
02-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

2 μm post- manufacturing filtered. This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Specifications

Specifications

Podocalyxin
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O00592
Podocalyxin-like protein 1 antibody, podolyxin, PCLP antibody, PCLP1 antibody, GCTM-2 antigen antibody.
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
B34D1.3
PE
PODXL
Liquid
PODXL
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
5420
Videos
Documents

For Research Use Only.