missing translation for 'onlineSavingsMsg'
Learn More

Podocalyxin Mouse anti-Human, PE, Clone: B34D1.3, eBioscience™

Mouse Monoclonal Antibody

£79.25 - £160.00

Specifications

Antigen Podocalyxin
Clone B34D1.3
Target Species Human
Conjugate PE
Isotype IgG2a, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15597136
View Documents
Affymetrix eBioscience
12-8873-41
25 Tests £79.25
25 tests View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
15507146
View Documents
Affymetrix eBioscience
12-8873-42
100 Tests £160.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2 μm post- manufacturing filtered. This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Specifications

Specifications

Podocalyxin
Human
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
B34D1.3
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.