Learn More
PNU 74654, Tocris Bioscience™
β-catenin binder; inhibits Wnt signaling
Brand:  Tocris (Bio-Techne) 353450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13463154
  • £527.73 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.
Specifications
Chemical Identifiers
113906-27-7
320.348
benzoic acid, 2-phenoxy-, 2-5-methyl-2-furanyl methylene hydrazide
N-[(Z)-(5-methylfuran-2-yl)methylideneamino]-2-phenoxybenzamide
C19H16N2O3
JJEDWBQZCRESJL-MOSHPQCFSA-N
68672283
CC1=CC=C(O1)C=NNC(=O)C2=CC=CC=C2OC3=CC=CC=C3
Specifications
50mg
113906-27-7
benzoic acid, 2-phenoxy-, 2-5-methyl-2-furanyl methylene hydrazide
CC1=CC=C(O1)C=NNC(=O)C2=CC=CC=C2OC3=CC=CC=C3
320.348
320.34
PNU 74654
C19H16N2O3
JJEDWBQZCRESJL-MOSHPQCFSA-N
N-[(Z)-(5-methylfuran-2-yl)methylideneamino]-2-phenoxybenzamide
68672283
>99%