missing translation for 'onlineSavingsMsg'
Learn More

PNU 37883 hydrochloride, Tocris Bioscience™

Vascular Kir6 (KATP) channel blocker

Brand:  Tocris (Bio-Techne) 209510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13430464

  • £169.05 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Novel antagonist selective for the vascular form of Kir6 (KATP) channel; inhibits Kir6 currents in isolated mesenteric artery smooth muscle cells (Kd = 65 nM) but not in cardiac or skeletal myocytes. Inhibits blood vessel relaxation and hypotension induced by pinacidil in vivo. Displays weak diuretic effects.
Specifications

Chemical Identifiers

57568-80-6
381.989
pnu 37883 hydrochloride, 4-morpholinecarboximidine-n-1-adamantyl-n'-cyclohexane, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-4-morpholinecarboximidamide monohydrochloride, 4-morpholinecarboximidamide, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-, monohydrochloride, c21h35n3o.hcl, n-1-adamantyl-n'-cyclohexylmorpholine-4-carboximidamide hydrochloride, n-cyclohexyl-n'-tricyclo 3.3.1.13,7 dec-1-yl-4-morpholinecarboximidamide hydrochloride, n-1-adamantyl-n inverted exclamation marka-cyclohexyl-4-morpholinecarboxamidine hydrochloride
N-(1-adamantyl)-N'-cyclohexylmorpholine-4-carboximidamide;hydrochloride
C21H36ClN3O
FZALCKUJYZCDOX-UHFFFAOYSA-N
64392
C1CCC(CC1)N=C(NC23CC4CC(C2)CC(C4)C3)N5CCOCC5.Cl

Specifications

10mg
57568-80-6
pnu 37883 hydrochloride, 4-morpholinecarboximidine-n-1-adamantyl-n'-cyclohexane, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-4-morpholinecarboximidamide monohydrochloride, 4-morpholinecarboximidamide, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-, monohydrochloride, c21h35n3o.hcl, n-1-adamantyl-n'-cyclohexylmorpholine-4-carboximidamide hydrochloride, n-cyclohexyl-n'-tricyclo 3.3.1.13,7 dec-1-yl-4-morpholinecarboximidamide hydrochloride, n-1-adamantyl-n inverted exclamation marka-cyclohexyl-4-morpholinecarboxamidine hydrochloride
C1CCC(CC1)N=C(NC23CC4CC(C2)CC(C4)C3)N5CCOCC5.Cl
381.989
381.98
PNU 37883 hydrochloride
C21H36ClN3O
FZALCKUJYZCDOX-UHFFFAOYSA-N
N-(1-adamantyl)-N'-cyclohexylmorpholine-4-carboximidamide;hydrochloride
64392
>99%
Videos
Documents