missing translation for 'onlineSavingsMsg'
Learn More

PNU 282987, Tocris Bioscience™

Selective α7 nAChR agonist

Brand:  Tocris (Bio-Techne) 230350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460984

  • £474.52 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Highly selective α7 nAChR agonist (Ki = 26 nM) displaying negligible blockade of α1β1γδ and α3β4 nAChRs (IC50 ≥ 60 μM). Found to be inactive against a panel of 32 receptors at 1 μM, except 5-HT3 receptors (Ki = 930 nM).
Specifications

Chemical Identifiers

123464-89-1
319.226
pnu-282987 hydrate, unii-916d7sp4cx, n-3r-1-azabicyclo 2.2.2 oct-3-yl-4-chloro-benzamide monohydrochloride hydrate, pnu-282987 monohydrate, n-3r-1-azabicyclo 2.2.2 octan-3-yl-4-chlorobenzamide hydrate hydrochloride, c14h17cln2o.clh.h2o, benzamide, n-3r-1-azabicyclo 2.2.2 oct-3-yl-4-chloro-, hydrochloride, hydrate 1:1:1, pnu-282987 hydrate, solid hplc
N-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-4-chlorobenzamide;hydrate;hydrochloride
C14H20Cl2N2O2
OCWLMMBVXIESNP-GXKRWWSZSA-N
16219883
C1CN2CCC1C(C2)NC(=O)C3=CC=C(C=C3)Cl.O.Cl

Specifications

PNU 282987
123464-89-1
pnu-282987 hydrate, unii-916d7sp4cx, n-3r-1-azabicyclo 2.2.2 oct-3-yl-4-chloro-benzamide monohydrochloride hydrate, pnu-282987 monohydrate, n-3r-1-azabicyclo 2.2.2 octan-3-yl-4-chlorobenzamide hydrate hydrochloride, c14h17cln2o.clh.h2o, benzamide, n-3r-1-azabicyclo 2.2.2 oct-3-yl-4-chloro-, hydrochloride, hydrate 1:1:1, pnu-282987 hydrate, solid hplc
C1CN2CCC1C(C2)NC(=O)C3=CC=C(C=C3)Cl.O.Cl
319.226
264.75
50mg
C14H20Cl2N2O2
OCWLMMBVXIESNP-GXKRWWSZSA-N
N-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-4-chlorobenzamide;hydrate;hydrochloride
16219883
>98%
Videos