Learn More
PNU 177864 hydrochloride, Tocris Bioscience™
Highly selective D3 antagonist
Brand:  Tocris (Bio-Techne) 335550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13432874
  • £605.64 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Highly selective dopamine D3 receptor antagonist. Induces phospholipidosis and exhibits antischizophrenic activity in vivo.
Specifications
Chemical Identifiers
250266-51-4
438.89
pnu 177864 hydrochloride, n-4-2-propylamino ethyl phenyl-4-trifluoromethoxy-benzenesulfonamide hydrochloride
N-[4-[2-(propylamino)ethyl]phenyl]-4-(trifluoromethoxy)benzenesulfonamide;hydrochloride
C18H22ClF3N2O3S
UOMDLHQTLPWCCE-UHFFFAOYSA-N
56972219
CCCNCCC1=CC=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)OC(F)(F)F.Cl
Specifications
50mg
250266-51-4
pnu 177864 hydrochloride, n-4-2-propylamino ethyl phenyl-4-trifluoromethoxy-benzenesulfonamide hydrochloride
CCCNCCC1=CC=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)OC(F)(F)F.Cl
438.89
438.89
PNU 177864 hydrochloride
C18H22ClF3N2O3S
UOMDLHQTLPWCCE-UHFFFAOYSA-N
N-[4-[2-(propylamino)ethyl]phenyl]-4-(trifluoromethoxy)benzenesulfonamide;hydrochloride
56972219
>99%