missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPLA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562185
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LRFLERRGLT KEPVLWTLVS KEPPAPADGN WDAGCDQRWK GGLSLNWKVP HVQVKDVPNF EQ Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 63%.
PNPLA5 Polyclonal antibody specifically detects PNPLA5 in Human samples. It is validated for Immunohistochemistry.Specifications
PNPLA5 | |
Polyclonal | |
Unconjugated | |
PNPLA5 | |
dJ388M5; dJ388M5.4; GS2L; GS2-like protein; patatin like phospholipase domain containing 5; patatin-like phospholipase domain containing 5; patatin-like phospholipase domain-containing protein 5; PNPLA5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150379 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6Z6 | |
PNPLA5 | |
Recombinant protein corresponding to Human PNPLA5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only