missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PMP Griffin Beaker

Make your job easier and safer while handling a wide range of liquids with these polymethylpentene beakers. VITLAB™ PMP Griffin Beakers feature an easy to read, embossed red scale.

Brand:  VITLAB™ 61403

Code : 02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12733409

  • £67.80 / Pack of 6


Estimated Shipment
18-06-2018
Add to basket

Description

Description

  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C
Specifications

Specifications

Yes
Crystal Clear
Yes
Printed Red Scale
Griffin Beaker
PMP (Polymethylpentene)
According to ISO 7056
For volume measurement
147mm
Griffin
1000mL
Videos