Learn More
PMF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567206
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MTQQIYDKFIA QLQTSIREEI SDIKEEGNLE AVLNALDKIV EEGKVRKEPA WRPSGIPEKD LHSVMAP Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.
PMF1 Polyclonal antibody specifically detects PMF1 in Human samples. It is validated for Immunocytochemistry.Specifications
PMF1 | |
Polyclonal | |
Unconjugated | |
PMF1 | |
2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P1K2 | |
PMF1 | |
Recombinant Human PMF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only