Learn More
PLZF Monoclonal Antibody (Mags.21F7), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£106.00 - £261.00
Specifications
Antigen | PLZF |
---|---|
Clone | Mags.21F7 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570267
|
Affymetrix eBioscience
53-9320-80 |
25 μg |
£106.00
25µg |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15580267
|
Affymetrix eBioscience
53-9320-82 |
100 μg |
£261.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This Mags.21F7 monoclonal antibody reacts with human and mouse promyelocytic leukemia zinc finger (PLZF), a member of the BTB-POZ family of transcription factors. Expression of this transcriptional repressor in immune cells differs between mice and humans. In mice, PLZF is highly expressed in immature CD1d-resricted NKT2 and NKT1 cells, and a subset of gamma delta (Vg1.1+Vd6.3+) T cells. Studies have also demonstrated expression of PLZF in non-invariant CD1d-restricted T cells, as well as non-CD1d-restricted innate T cells. In humans, PLZF is expressed in NK cells, gamma delta T cells, as well as CD4 and CD8+ T cells. PLZF is also expressed in MR1-specific mucosal-associated invariant T cells, as well as in MHC Class II-restricted T cells that develop via a thymocyte-thymocyte interaction in humans. PLZF exists as a homodimer or in complex with PLZP, and has been shown to be involved in the development of NKT cells, NK cell function, cellular quiescence, and growth suppression. Finally, PLZF has been shown to inhibit gene expression induced by retinoic acid receptor. Applications Reported: This Mags.21F7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This Mags.21F7 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 1 μg per test.
PLZF Monoclonal antibody specifically detects PLZF in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
PLZF | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
ZBTB16 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ZBTB16 |
Mags.21F7 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human, Mouse | |
Q05516 | |
235320, 7704 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.