missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PLUNC Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | PLUNC |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
PLUNC Polyclonal specifically detects PLUNC in Human samples. It is validated for Western Blot, ELISA.Specifications
PLUNC | |
Western Blot 0.1 ug/mL, ELISA | |
Polyclonal | |
Goat | |
Human | |
bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl | |
BPIFA1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Q9NP55 | |
51297 | |
E. coli-derived recombinant human PLUNC Gln20-Val256 Accession # Q9NP55 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human PLUNC in direct ELISAs and Western blots. |
For Research Use Only