missing translation for 'onlineSavingsMsg'
Learn More

Plexin-B2, clone: eBio3E7 (3E7), eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5665-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247427

  • £68.21 / 50µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved coverting the single chain presursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2's possible role in the immune system has not been characterized.Applications Reported: This eBio3E7 (3E7) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining.Applications Tested: This eBio3E7 (3E7) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Plexin-B2
eBio3E7 (3E7)
Unconjugated
Purified
Armenian Hamster
50μg
Store at 2-8°C.
Monoclonal
Immunoprecipitation
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PlexinB2
IgG
RUO
Primary
Murine
Videos