Learn More
PLEKHG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555582
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CQDHLYNSLG RKGISAKSQP YHRSQSSSSV LINKSMDSIN YPSDVGKQQL LSLHRSSRCE SHQDLLPDIA DSHQQGTEKL SDLTLQDSQK VVVVN Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 74%.
PLEKHG1 Polyclonal antibody specifically detects PLEKHG1 in Human samples. It is validated for Immunohistochemistry.Specifications
PLEKHG1 | |
Polyclonal | |
Unconjugated | |
PLEKHG1 | |
ARHGEF41; KIAA1209; pleckstrin homology and RhoGEF domain containing G1; pleckstrin homology domain containing, family G (with RhoGef domain) member 1; pleckstrin homology domain-containing family G member 1; PLEKHG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULL1 | |
PLEKHG1 | |
Recombinant protein corresponding to Human PLEKHG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only