missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, Hard, 99.95% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 014195.BW

Code : NEW

Additional Details : CAS Number : 7440-06-4 Weight : 0.01000kg

Quantity 50cm

 View more versions of this product

Product Code. 11317787

  • £576.60 / 50cm


Estimated Shipment date
from Supplier 23-04-2018
Add to basket

Specifications

Chemical Identifiers

Platinum wire
∽9.78g/m, Temper: As Drawn
MFCD00011179
7440-06-4
99.95% (metals basis)

Specifications

Platinum wire
7440-06-4
99.95% (metals basis)
50cm
∽9.78g/m, Temper: As Drawn
MFCD00011179
Videos
SDS