missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum Ultrathin foil, 0.1 micron thick, 99.99% (metals basis)

Brand:  Alfa Aesar™ 040699.DH

Code : NEW

Additional Details : CAS Number : 7440-06-4 Weight : 0.01000kg

Quantity 25mm

 View more versions of this product

Product Code. 11308638

  • £451.77 / Each


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Platinum Ultrathin foil
7440-06-4
MFCD00011179

Specifications

Platinum Ultrathin foil
7440-06-4
99.99%
MFCD00011179
0.1 Micron thick
25mm
Circular Disc on Permanent Mylar Support
(metals basis)
Odorless
Videos
SDS