missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Premion™, 99.99+% (metals basis), Thermo Scientific Chemicals
684.03 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 Pc.
Description
Platinum slug finds application in jewelry production, electrical contacts for semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 7440-06-4 |
Molecular Formula | Pt |
MDL Number | MFCD00011179 |
Quantity | 1 Pc. |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
Solubility Information | Insoluble in water. |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
SMILES | [Pt] |
IUPAC Name | platinum |
Molecular Weight (g/mol) | 195.08 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'