missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Platinum Rhodium foil, 0.2mm (0.008 in.) thick, 99.9% (metals basis)

Brand:  Thermo Scientific Alfa Aesar 042837.FF

Platinum Rhodium foil, 0.2mm (0.008 in.) thick, 99.9% (metals basis)

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11340048

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.