missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum Rhodium foil, 0.2mm (0.008 in.) thick, 99.9% (metals basis)

Brand:  Alfa Aesar™ 042837.FI

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50x50mm

 View more versions of this product

Product Code. 11366159

  • £99999.00 / Each


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Platinum Rhodium foil
0.2mm (0.008 in.) thick
MFCD00198186

Specifications

Pt:Rh; 80:20 wt%
≈2.3g/25x25mm
0.2mm (0.008 in.) thick
99.9%
MFCD00198186
Platinum Rhodium foil
50x50mm
Foil
(metals basis)
Odorless
Videos
SDS