Learn More
Platinum Rhodium foil, 0.025mm (0.001 in.) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041808.GH
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Rhodium foil | |
Pt:Rh; 90:10 wt% | |
MFCD00198186 | |
Insoluble in water. | |
Odorless | |
Foil |
0.025mm (0.001 in.) thick | |
(metals basis) | |
100 x 100 mm | |
99.9% | |
≈0.31g/25x25mm |
RUO – Research Use Only