missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum powder, amorphous, <10 micron, 99.9% (metals basis)

Brand:  Alfa Aesar™ 012100.77

Code : NEW

Additional Details : CAS Number : 7440-06-4 Weight : 0.00010kg

Quantity 0.1g

 View more versions of this product

Product Code. 11366458

  • £27.50 / 0.10g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Platinum powder
7440-06-4
MFCD00011179

Specifications

Platinum powder
Amorphous Powder
(metals basis)
Odorless
9.2 to 15.7m2/g
Tap density: 0.9g/cm3 to 1.5g/cm3
7440-06-4
99.9%
MFCD00011179
<10 Micron
0.1g
Videos
SDS