missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 5% on alumina powder, reduced, Alfa Aesar™

Brand:  Alfa Aesar™ 044222.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461977

  • £73.00 / 5g


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Specifications

Chemical Identifiers

Powder
MFCD00011179
Powder
Platinum
5% on alumina powder

Specifications

Platinum
5% on alumina powder
Powder
MFCD00011179
5g
7440-6-6
Powder
Videos
SDS