missing translation for 'onlineSavingsMsg'
Learn More

Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number 11302328
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
7440-6-6
Pt in 20% HCl
MFCD00011179
This item is not returnable. View return policy
7440-6-6
Pt in 20% HCl
MFCD00011179

Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Odor Characteristic
Density Approx. 1g/cm3
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Platinum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'