missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, nominally 40%, Ruthenium nominally 20% on 50% compressed carbon black, HiSPEC™ 10300, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 1 g |
UN Number | UN1325 |
Chemical Name or Material | Platinum, Nominally 40%, Ruthenium nominally 20% on 50% compressed carbon black, HiSPEC™ 10300 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'