missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, nominally 20% on carbon black, HiSPEC™ 3000, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Gray to Black |
Odor | Odorless |
Quantity | 1 g |
UN Number | UN1325 |
Physical Form | Powder |
Chemical Name or Material | Platinum, nominally 20% on carbon black, HiSPEC™ 3000 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'