missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Platinum(IV) oxide hydrate is used in the conversion of ketones to methylenes by hydrogenolysis of enol triflates under neutral conditions. It is used as a catalyst for hydrosilylation of olefins with various alkyl dialkoxy silanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Physical Form | Powder |
Quantity | 5 g |
Linear Formula | PtO2·xH2O |
UN Number | UN1479 |
Merck Index | 14,7527 |
Solubility Information | Insoluble in water. Soluble in concentrated acids and dilute solution of potassium hydroxide. Slightly soluble in acetone and chloroform. |
Formula Weight | 227.09 (Anhydrous) |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | Pt 71 to 75% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'