missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide, 83% Pt, Thermo Scientific Chemicals

Catalog Number 10563941
101.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1314-15-4
O2Pt
227.08
MFCD00011184
YKIOKAURTKXMSB-UHFFFAOYSA-N
Adams' catalyst
dioxoplatinum
O=[Pt]=O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1314-15-4
O2Pt
227.08
MFCD00011184
YKIOKAURTKXMSB-UHFFFAOYSA-N
Adams' catalyst
dioxoplatinum
O=[Pt]=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 450.0°C
Color Brown
Physical Form Powder
Quantity 1 g
Linear Formula PtO2
UN Number 1479
Merck Index 15, 7642
Solubility Information Solubility in water: insoluble
Formula Weight 227.09
Percent Purity 83% (Pt)
Packaging Glass bottle
Chemical Name or Material Platinum(IV) oxide, 83% Pt
Show More Show Less
compliance-icons
Product Identifier
  • Platinum(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'