missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407698
Click to view available options
Quantity:
10 cm
50 cm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

Form: approx. 0.11g/10cm,; Temper: soft

Used in the fabrication of metal microelectrodes for electrical stimulation of nervous tissues and in electrophysiological recordings. Inoculating needles and leadwires for implants. Used in electrode materials, composing the gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver gray wire
Assay Percent Range >99.9%
Quantity 10 cm
Solubility Information Insoluble in water.
Odor Odorless
Form Wire
Chemical Name or Material Platinum Iridium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'