missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Platinum Iridium wire, 1.0mm (0.04 in.) dia.
Brand: Alfa Aesar™ 042678.BY
Code : NEW
Additional Details : Weight : 0.01000kg
Quantity | 100cm |
---|
Chemical Identifiers
Platinum Iridium wire | |
1.0mm (0.04 in.) dia. | |
MFCD02091683 |
Specifications
Pt:Ir; 90:10 wt% | |
≈0.85g/5cm | |
1.0mm (0.04 in.) dia. | |
MFCD02091683 |
Platinum Iridium wire | |
100cm | |
Wire | |
Odorless |