missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 0.5% on granular carbon, reduced, Thermo Scientific Chemicals

Catalog Number p-4782065
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-6-6
MFCD00011179
This item is not returnable. View return policy
7440-6-6
MFCD00011179

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Appearance Black granules
Quantity 5 g
Solubility Information Soluble in aqua regia. Insoluble in water,mineral and organic acids.
Odor Odorless
Form Granular
Chemical Name or Material Platinum, 0.5% on granular carbon

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'