Learn More
Cole-Parmer™ Platinum-cured Silicone Seal Tri-Clamp Gaskets Fitting
Brand:  Cole-Parmer™ 30523-80
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11786613
  • £78.90 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Specifications
12.7 mm dia.
Strainer Seal Tri-Clamp
Platinum-cured Silicone
For pharmaceutical, high-purity water, injectable, food, and beverage applications
1 Each