missing translation for 'onlineSavingsMsg'
Learn More

Platinum on activated carbon, 1% Pt, ACROS Organics™

Brand:  Acros Organics 195220250

Code : 29

Additional Details : CAS Number : 7440-06-4 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10636592

  • £67.60 / 25g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Platinum on activated carbon
7440-06-4
Pt
MFCD00011179
1% Pt
1% (Pt)
195.08

Specifications

Platinum on activated carbon
1% Pt
Powder
Pt
MFCD00011179
Typical particle size distribution: D50: 28 Micron, <8 Micron: 14%, <16 Micron: 29%, <32 Micron: 55%, <64 Micron: 84%, <128 Micron: 99%
25g
7440-06-4
1% (Pt)
195.08
Glass bottle
Black
Videos
SDS