missing translation for 'onlineSavingsMsg'
Learn More

Platinum, AAS standard solution, Pt 1000μg/mL, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 088086.AP

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1789 Chem class : 1a Pack group : II

Quantity 500mL

 View more versions of this product

Product Code. 11306369

  • £191.30 / 500mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Platinum
Pt 1000μg/mL
MFCD00011179
Liquid
Pt in 20% HCl

Specifications

Platinum
Pt 1000μg/mL
MFCD00011179
500mL
Liquid
Pt in 20% HCl
Characteristic
Videos
SDS