Learn More
Platinum 6 wt% Rhodium wire, 0.33mm (0.013in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 043025.G2
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 6 wt% Rhodium wire | |
Pt:Rh; 94:6 wt% | |
2 m | |
≈0.88g/50cm |
0.33mm (0.013 in.) dia., ISA Type B Standard Grade Thermocouple | |
MFCD00198186 | |
Odorless | |
Wire |
RUO – Research Use Only