missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 5% on activated carbon paste, Type

Brand:  Alfa Aesar™ H37641.14

Code : NEW

Additional Details : CAS Number : 7440-06-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15409105

  • £191.50 / 25g


Estimated Shipment date
from Supplier 20-06-2018
Add to basket

Specifications

Chemical Identifiers

Platinum
7440-06-4
5% on activated carbon paste, Type 128M
MFCD00011179

Specifications

Platinum
7440-06-4
MFCD00011179
Black
5% on activated carbon paste, Type 128M
Solid
Odorless
25g
Videos
SDS