Learn More
Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
For ISA Type B Standard Grade Thermocouple
Brand: Thermo Scientific Alfa Aesar 012576.BY
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Platinum 30 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple is used in the glass industry for furnace linings and complex fabrications. It is used as a catalyst in the preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 30 wt% Rhodium wire | |
Wire | |
MFCD00198186 | |
Odorless |
0.5mm (0.02 in.) dia. | |
100 cm | |
Insoluble in water. |
RUO – Research Use Only