missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type R Standard Grade Thermocouple

For ISA Type R Standard Grade Thermocouple

Brand:  Alfa Aesar™ 010062.BU

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25cm

 View more versions of this product

Product Code. 11356767

  • £73.60 / 25cm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Platinum 13 wt% Rhodium wire
0.25mm (0.01 in.) dia.
MFCD00198186

Specifications

Pt:Rh; 87:13 wt%
≈0.26g/25cm
0.25mm (0.01 in.) dia.
MFCD00198186
Platinum 13 wt% Rhodium wire
25cm
Wire
Odorless
Videos
SDS